Learn More
R&D Systems™ Recombinant Human Kallikrein 5 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand:  R&D Systems™ 1108-SE-010
454.42 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18310608
  • 595.00 € / 10µg
Explore more special offers
Description
Description
The Recombinant Human Kallikrein 5 Protein is derived from NS0. The Recombinant Human Kallikrein 5 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in MES, NaCl and Glycerol.
31 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
25818
Kallikrein 5
Mouse myeloma cell line,NS0-derived human Kallikrein 5 protein Ile67-Ser293,with a C-terminal 10-His tag
Mature form
Recombinant
Product Suggestions