missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Kallikrein 7 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 2624-SE-010
394.81 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human Kallikrein 7 Protein is derived from NS0. The Recombinant Human Kallikrein 7 Protein has been validated for the following applications: Enzyme Activity.Specifications
Supplied as a 0.2μm filtered solution in HEPES and NaCl. | |
26 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
5650 | |
Kallikrein 7 | |
Mouse myeloma cell line,NS0-derived human Kallikrein 7 protein Glu23-His252,with a C-terminal 10-His tag | |
Pro form | |
Recombinant |