Learn More
R&D Systems™ Recombinant Human LRP-1 Cluster IV Fc Chimera Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand:  R&D Systems™ 5395-L4-050
429.36 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18107010
  • 561.00 € / 50µg
Explore more special offers
Description
Description
Measured by its binding ability in a functional ELISA. In a Human IgG Fc Antibody (Catalog # G-102-C) coated plate, Recombinant Human LRP-1 Cluster IV Fc Chimera binds Recombinant Human LRPAP (Catalog # 4296-LR ) with an ED50 of 0.25-1.25 ng/mL.
Specifications
Specifications
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
76.7 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
4035
LRP-1 Cluster IV Fc Chimera
Chinese Hamster Ovary cell line,CHO-derived human LRP-1 Cluster IV protein, (N-terminus) Human LRP1-C4 (Ser3332-Asp3779) Accession &Num; Q07954 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Disulfide-linked homodimer
Recombinant
Product Suggestions