missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LYVE1 Protein

A cDNA sequence encoding the LYVE1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12609-10ug

 View more versions of this product

Product Code. 15960979

  • 203.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

10894
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Baculovirus
DPLRAEELS IQVSCRIMGI TLVSKKANQQ LNFTEAKEAC RLLGLSLAGK DQVETALKAS FETCSYGWVG DGFVVISRIS PNPKCGKNGV GVLIRKVPVS RQFAAYCYNS SDTWTNSCIP EIITTKDPIF NTQTATQTTE FIVSDSTYSV ASPYSTIPAP TTTPPAPAST SIPRRKKLIC VTEVFMETST MSTETEPFVE NKAAFKNEAA GFGGSGRLVP RGSHHHHHH
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
LYVE1 Protein
Research Use Only
LYVE1
Human
Untagged
The LYVE1 protein solution contains 20mM Tris buffer pH 7.5 and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers