missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human NTB-A/SLAMF6 Fc Chimera Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 1908-NT-050

272.13 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18187410

  • 356.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human NTB-A/SLAMF6 Fc Chimera Protein is derived from NS0. The Recombinant Human NTB-A/SLAMF6 Fc Chimera Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
M.W.-observed-55-80 kDa, reducing conditions
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
114836
NTB-A/SLAMF6 Fc Chimera
Mouse myeloma cell line,NS0-derived human NTB-A/SLAMF6 protein, (N-terminus) Human NTB-A (Leu28-Lys225) Accession &Num; Q96DU3 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Disulfide-linked homodimer
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers