Learn More
R&D Systems™ Recombinant Human OSMR beta Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand:  R&D Systems™ 4389-OR-050
Product Code. 18127250
  • 367.00 € / 50µg
Explore more special offers
Description
Description
The Recombinant Human OSM R beta Protein is derived from NS0. The Recombinant Human OSM R beta Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
81 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
9180
OSMR beta
Mouse myeloma cell line,NS0-derived human OSM R beta protein Glu28-Ser739
IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSM R beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1
Recombinant
Product Suggestions