Learn More
R&D Systems™ Recombinant Human PA28 Activator alpha Subunit Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ E-381-100
257.45 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 30001741
  • 332.00 € / 100µg
Explore more special offers
Description
Description
The Recombinant Human PA28 Activator alpha Subunit Protein is derived from E. coli. The Recombinant Human PA28 Activator alpha Subunit Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
100 μg
Human
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.