Learn More
R&D Systems™ Recombinant Human Sirtuin 3/SIRT3 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand:  R&D Systems™ 7488-DA-050
Product Code. 18148470
  • 373.00 € / 50µg
Explore more special offers
Description
Description
The Recombinant Human Sirtuin 3/SIRT3 Protein is derived from E. coli. The Recombinant Human Sirtuin 3/SIRT3 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in Tris, NaCl, Brij-35 and Glycerol.
34 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Recombinant
23410
Sirtuin 3/SIRT3
E. coli-derived human Sirtuin 3/SIRT3 protein Ser101-Lys399,with an N-terminal Met and 6-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane.