Learn More
R&D Systems™ Recombinant Human SorLA (aa 82-753) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 9880-LA-050
394.81 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18158530
  • 509.00 € / 50µg
Explore more special offers
Description
Description
The Recombinant Human SorLA (aa 82-753) Protein is derived from HEK293. The Recombinant Human SorLA (aa 82-753) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Q92673
Lyophilized from a 0.2 μm filtered solution in PBS.
76 kDa
Human embryonic kidney cell, HEK293-derived human SorLA protein Ser82-Pro753, with a C-terminal 6-His tag
RUO
Unconjugated
Human
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Bioactivity
6653
50 μg
12 months from date of receipt, ≤ -20°C as supplied; 1 month, 2 to 8°C under sterile conditions after reconstitution; 3 months, ≤ -20°C under sterile conditions after reconstitution.
<0.10 EU per 1 μg of the protein by the LAL method.
SorLA
Reconstitute at 1 mg/mL in PBS.