Learn More
R&D Systems™ Recombinant Human Sortilin Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand:  R&D Systems™ 3154-ST-050
433.68 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18367767
  • 570.00 € / 50µg
Explore more special offers
Description
Description
The Recombinant Human Sortilin Protein is derived from NS0. The Recombinant Human Sortilin Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
76.6 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
6272
Sortilin
Mouse myeloma cell line,NS0-derived human Sortilin protein Ser78-Asn755,with a C-terminal 6-His tag
Monomer
Recombinant
Product Suggestions