Learn More
R&D Systems™ Recombinant Human Sphingosine Kinase 1/SPHK1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand:  R&D Systems™ 5536-SK-010
Product Code. 18187580
  • 347.00 € / 10µg
Explore more special offers
Description
Description
The Recombinant Human Sphingosine Kinase 1/SPHK1 Protein is derived from Sf 21 (baculovirus). The Recombinant Human Sphingosine Kinase 1/SPHK1 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2 μm filtered solution in MES, NaCl, Glycerol, DTT and Triton™ .
45 kDa
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Recombinant
8877
Sphingosine Kinase 1/SPHK1
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Sphingosine Kinase 1/SPHK1 protein Asp2-Leu398 with an N-terminal Met and 6-His tag
Unconjugated
>75%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.