missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Steroid 17-alpha-hydroxylase / 17,20 lyase Protein
A DNA sequence encoding the Homo sapiens (Human) Steroid 17-alpha-hydroxylase/17,20 lyase, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_000093.1 |
---|---|
Gene ID (Entrez) | 1586 |
Name | Steroid 17-alpha-hydroxylase / 17,20 lyase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948569
|
enQuireBio™
QP5905-ec-10ug |
10 μg | |||||||
15988569
|
enQuireBio™
QP5905-ec-50ug |
50 μg | |||||||
15938569
|
enQuireBio™
QP5905-ec-100ug |
100 μg |
505.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15968569
|
enQuireBio™
QP5905-ec-200ug |
200 μg | |||||||
15978569
|
enQuireBio™
QP5905-ec-500ug |
500 μg | |||||||
15958569
|
enQuireBio™
QP5905-ec-1mg |
1 mg |
2285.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_000093.1 | |
Steroid 17-alpha-hydroxylase / 17,20 lyase Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1586 | |
Research Use Only | |
CYP17A1 | |
Recombinant Protein | |
E. coli | |
GAKYPKSLLS LPLVGSLPFL PRHGHMHNNF FKLQKKYGPI YSVRMGTKTT VIVGHHQLAK EVLIKKGKDF SGRPQMATLD IASNNRKGIA FADSGAHWQL HRRLAMATFA LFKDGDQKLE KIICQEISTL CDMLATHNGQ SIDISFPVFV AVTNVISLIC FNTSYKNGDP ELNVIQNYNE GIIDNLSKDS LVDLVPWLKI FPNKTLEKLK SHVKIRNDLL NKILENYKEK FRSDSITNML DTLMQAKMNS DNGNAGPDQD SELLSDNHIL TTIGDIFGAG VETTTSVVKW TLAFLLHNPQ VKKKLYEEID QNVGFSRTPT ISDRNRLLLL EATIREVLRL RPVAPMLIPH KANVDSSIGE FAVDKGTEVI INLWALHHNE KEWHQPDQFM PERFLNPAGT QLISPSVSYL PFGAGPRSCI GEILARQELF LIMAWLLQRF DLEVPDDGQL PSLEGIPKVV FLIDSFKVKI KVRQAWREAQ AEGST | |
Greater than 80% as determined by SDS-PAGE. |