missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human sTNF RI/TNFRSF1A Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 636-R1-025
425.90 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human sTNF RI/TNFRSF1A Protein is derived from E. coli. The Recombinant Human sTNF RI/TNFRSF1A Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
21 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
7132 | |
TNF RI/TNFRSF1A | |
E. coli-derived human TNF RI/TNFRSF1A protein Ile22-Thr211,with an N-terminal Met | |
Unconjugated | |
Reconstitute at 100μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin. |