missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human THAP3 Protein
A cDNA sequence encoding the THAP3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13740-1mg
This item is not returnable.
View return policy
Specifications
THAP3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPKSCAA RQCCNRYSSR RKQLTFHRFP FSRPELLKEW VLNIGRGNFK PKQHTVICSE HFRPECFSAF GNRKNLKHNA VPTVFAFQDP TQQVRENTDP ASERGNASSS QKEKVLPEAG AGEDSPGRNM DTALEELQLP PNAEGHVKQV SPRRPQATEA VGRPTGPAGL RRTPNKQPSD HSYALLDLDS LKKKLFLTLK ENEKLRKRLQ AQRLVMRRMS SRLRACKGHQ GLQARLGPEQ QS | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The THAP3 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 2mM DTT and 50% glycerol. |