missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human TIMP-4 (Sf 21-expressed) Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity

Brand:  R&D Systems™ 974-TSF-010

314.46 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18177900

  • 411.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human TIMP-4 (Sf 21-expressed) Protein is derived from Sf 21 (baculovirus). The Recombinant Human TIMP-4 (Sf 21-expressed) Protein has been validated for the following applications: Inhibition Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Tris and NaCl.
22 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
7079
TIMP-4
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human TIMP-4 protein Cys30-Pro224
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers