Learn More
R&D Systems™ Recombinant Human TLR4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 1478-TR-050
Product Code. 18167200
  • 508.00 € / 50µg
Explore more special offers
Description
Description
The Recombinant Human TLR4 Protein is derived from NS0. The Recombinant Human TLR4 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
70.6 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
7099
TLR4
Mouse myeloma cell line,NS0-derived human TLR4 protein Glu24-Lys631,with a C-terminal Ser and 10-His tag
<0.1 EU per 1μg of the protein by the LAL method.
Recombinant
Product Suggestions