Learn More
R&D Systems™ Recombinant Human TSG-6 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand:  R&D Systems™ 2104-TS-050
Product Code. 18197920
  • 591.00 € / 50µg
Explore more special offers
Description
Description
The Recombinant Human TSG-6 Protein is derived from NS0. The Recombinant Human TSG-6 Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
30.5 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
7130
TSG-6
Mouse myeloma cell line,NS0-derived human TSG-6 protein Trp18-Leu277,with a C-terminal 10-His tag
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.