missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TXNRD3NB Protein
A cDNA sequence encoding the TXNRD3NB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13843-0.1mg
This item is not returnable.
View return policy
Specifications
645840 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMRDLSER RLGQPELKAE QQMPLEPVRA RLSVGLACCC SHTTAEASSL EHGDKVFGQG FPSPLEEIKR LLKISRALQA RSVPSTQEKA KCLSGEPGQP EGKGQETYPG PGKVEGKAEP AMRKDDVCPG MKCISG | |
Greater than 85.0% as determined by SDS-PAGE. |
TXNRD3NB Protein | |
Research Use Only | |
TXNRD3NB | |
Human | |
His | |
TXNRD3NB protein solution (0.25 mg/ml) containing 20mM Tris-HCl (pH 8.0), 30% glycerol, 0.15M NaCl and 1mM DTT. |