missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBE2A Protein
A cDNA sequence encoding the UBE2A was constructed and used to recombinantly synthesize the protein.
131.00 € - 3275.00 €
Specifications
Name | UBE2A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15964909
|
enQuireBio™
QP13852-5ug |
5 μg |
131.00 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15954909
|
enQuireBio™
QP13852-25ug |
25 μg |
203.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15944909
|
enQuireBio™
QP13852-1mg |
1 mg |
3275.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
UBE2A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
UBE2A Human solution containing 20mM Tris HCl pH 8.0, 1mM DTT, 1mM EDTA and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSTPARRRLM RDFKRLQEDP PAGVSGAPSE NNIMVWNAVI FGPEGTPFED GTFKLTIEFT EEYPNKPPTV RFVSKMFHPN VYADGSICLD ILQNRWSPTY DVSSILTSIQ SLLDEPNPNS PANSQAAQLY QENKREYEKR VSAIVEQSWR DC | |
Greater than 95% as determined by SDS-PAGE. |