missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UBL5 Protein

A cDNA sequence encoding the UBL5 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13874-20ug

 View more versions of this product

Product Code. 15954979

  • 203.00 € / 20µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

UBL5 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MIEVVCNDRL GKKVRVKCNT DDTIGDLKKL IAAQTGTRWN KIVLKKWYTI FKDHVSLGDY EIHDGMNLEL YYQ
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
UBL5 protein solution (1 mg/1ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 2mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers