Learn More
R&D Systems™ Recombinant Human Uteroglobin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 4218-UT-050
346.42 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18108710
  • 440.00 € / 50µg
Explore more special offers
Description
Description
The Recombinant Human Uteroglobin Protein is derived from E. coli. The Recombinant Human Uteroglobin Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
8 kDa (monomer)
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
7356
Uteroglobin/SCGB1A1
E. coli-derived human Uteroglobin/SCGB1A1 protein Glu22-Asn91,with an N-terminal Met
Disulfide-linked homodimer
Recombinant
Product Suggestions