Learn More
R&D Systems™ Recombinant Human VEGF 111 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 5336-VE-010/CF
338.66 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18147850
  • 443.00 € / 10µg
Explore more special offers
Description
Description
The Recombinant Human VEGF 111 Protein is derived from E. coli. The Recombinant Human VEGF 111 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
13 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
7422
VEGF 111
E. coli-derived human VEGF protein Ala27-Arg137,with an N-terminal Met & Pro28-Arg137
Disulfide-linked homodimer
Recombinant
Product Suggestions