missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human VEGF 111 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

443.00 €

Specifications

Gene ID (Entrez) 7422
Molecular Weight (g/mol) 13 kDa
Name VEGF 111
Quantity 10 μg
Source E. coli-derived human VEGF protein Ala27-Arg137,with an N-terminal Met & Pro28-Arg137
View More Specs

Products 2
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
18147850
View Documents
R&D Systems™
5336-VE-010/CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
443.00 €
10µg
Please to purchase this item. Need a web account? Register with us today!
18376377
View Documents
R&D Systems™
5336-VE-010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
443.00 €
10µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Recombinant Human VEGF 111 Protein is derived from E. coli. The Recombinant Human VEGF 111 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

7422
VEGF 111
E. coli-derived human VEGF protein Ala27-Arg137,with an N-terminal Met & Pro28-Arg137
Disulfide-linked homodimer
Recombinant
13 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Videos
SDS
Documents

Documents

Product Certifications