missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human VEGF 121 (aa 207-327) Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

431.00 €

Specifications

Gene ID (Entrez) 7422
Name VEGF 121, aa 207-327
Quantity 10 μg
Source E. coli-derived human VEGF protein Ala207-Arg327,with an N-terminal Met & Pro208-Arg327
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
View More Specs

Products 2
Product Code Brand Formulation Molecular Weight (g/mol) Price Quantity & Availability  
Product Code Brand Formulation Molecular Weight (g/mol) Price Quantity & Availability  
18176130
View Documents
R&D Systems™
4644-VS-010
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. 14 kDa (monomer)
431.00 €
10µg
Please to purchase this item. Need a web account? Register with us today!
18106900
View Documents
R&D Systems™
4644-VS-010/CF
Lyophilized from a 0.2μm filtered solution in HCl. 14 kDa
431.00 €
10µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Recombinant Human VEGF 121 (aa 207-327) Protein is derived from E. coli. The Recombinant Human VEGF 121 (aa 207-327) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

7422
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
VEGF 121, aa 207-327
E. coli-derived human VEGF protein Ala207-Arg327,with an N-terminal Met & Pro208-Arg327
Disulfide-linked homodimer
Recombinant
Videos
SDS
Documents

Documents

Product Certifications