Learn More
R&D Systems™ Recombinant Human Vimentin Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand:  R&D Systems™ 2105-VI-100
387.90 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18702084
  • 507.00 € / 100µg
Explore more special offers
Description
Description
The Recombinant Human Vimentin Protein is derived from E. coli. The Recombinant Human Vimentin Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
54.4 kDa
100 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
7431
Vimentin
E. coli-derived human Vimentin protein Ser2-Glu466,with a C-terminal 6-His tag
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.