missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zinc finger protein GLI1 Protein

A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein GLI1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8750-ec-100ug

 View more versions of this product

Product Code. 16086281

  • 554.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001153517.1
Zinc finger protein GLI1 Protein
Research Use Only
GLI1
Recombinant Protein
E. coli
QEPSYQSPKF LGGSQVSPSR AKAPVNTYGP GFGPNLPNHK SGSYPTPSPC HENFVVGANR ASHRAAAPPR LLPPLPTCYG PLKVGGTNPS CGHPEVGRLG GGPALYPPPE GQVCNPLDSL DLDNTQLDFV AILDEPQGLS PPPSHDQRGS SGHTPPPSGP PNMAVGNMSV LLRSLPGETE FLNSSA
Greater than 90% as determined by SDS-PAGE.
2735
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers