missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZNF32 Protein
A cDNA sequence encoding the ZNF32 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP14003-5ug
This item is not returnable.
View return policy
Specifications
ZNF32 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMFGFPTA TLLDCHGRYA QNVAFFNVMT EAHHKYDHSE ATGSSSWDIQ NSFRREKLEQ KSPDSKTLQE DSPGVRQRVY ECQECGKSFR QKGSLTLHER IHTGQKPFEC THCGKSFRAK GNLVTHQRIH TGEKPYQCKE CGKSFSQRGS LAVHERLHTG QKPYECAICQ RSFRNQSNLA VHRRVHSGEK PYRCDQCGKA FSQKGSLIVH IRVHTGLKPY ACTQCRKSFH TRGNCILHGK IHTGETPYLC GQCGKSFTQR GSLAVHQRSC SQRLTL | |
Greater than 85.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ZNF32 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |