missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse BTC Protein
A cDNA sequence encoding the BTC was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 12223 |
---|---|
Name | BTC Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Btc |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974178
|
enQuireBio™
QP10539-5ug |
5 μg |
131.00 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15964178
|
enQuireBio™
QP10539-20ug |
20 μg |
203.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15954178
|
enQuireBio™
QP10539-1mg |
1 mg |
3915.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
12223 | |
Research Use Only | |
Btc | |
Mouse | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4. |
BTC Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
DGNTTRTPET NGSLCGAPGE NCTGTTPRQK VKTHFSRCPK QYKHYCIHGR CRFVVDEQTP SCICEKGYFG ARCERVDLFY | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |