missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Carbonic Anhydrase IV Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 2414-CA-010

234.12 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18394108

  • 302.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Mouse Carbonic Anhydrase IV Protein is derived from NS0. The Recombinant Mouse Carbonic Anhydrase IV Protein has been validated for the following applications: Enzyme Activity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in Tris and NaCl.
31 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Recombinant
12351
Carbonic Anhydrase IV/CA4
Mouse myeloma cell line,NS0-derived mouse Carbonic Anhydrase IV/CA4 protein Glu18-Ser277,with a C-terminal 10-His tag Accession &Num; Q64444
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers