Learn More
R&D Systems™ Recombinant Mouse Carboxyl Ester Lipase/CEL Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand:  R&D Systems™ 5658-CE-010
300.64 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18127480
  • 382.00 € / 10µg
Explore more special offers
Description
Description
The Recombinant Mouse Carboxyl Ester Lipase/CEL Protein is derived from NS0. The Recombinant Mouse Carboxyl Ester Lipase/CEL Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in Tris and NaCl.
58 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
12613
Carboxyl Ester Lipase/CEL
Mouse myeloma cell line,NS0-derived mouse Carboxyl Ester Lipase/CEL protein Ala21-Leu534,with a C-terminal 6-His tag
Monomer
Recombinant
Product Suggestions