Learn More
R&D Systems™ Recombinant Mouse CCL27/CTACK Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 725-CK-025/CF
 View more versions of this product
Product Code. 18359697
  • 253.00 € / 25µg
Explore more special offers
Description
Description
The Recombinant Mouse CCL27/CTACK Protein is derived from E. coli. The Recombinant Mouse CCL27/CTACK Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
10.9 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
20301
CCL27/CTACK
E. coli-derived mouse CCL27/CTACK protein Leu26-Asn120
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.