missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CCL4/MIP-1 beta Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 451-MB-050/CF

383.58 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


 View more versions of this product

Product Code. 18147110

  • 494.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CCR5. The ED50 for this effect is 0.75-5 ng/mL.
Specifications

Specifications

Q5QNV9
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
M.W. (theoretical): 7.8 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Ccl4
Recombinant
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Bioactivity
6351
CCL4/MIP-1 beta
E. coli-derived mouse CCL4/MIP-1 beta protein Ala24-Asn92
<0.01 EU / 1 μg of the protein by the LAL method.
Unconjugated
Reconstitute at 100 μg/mL in sterile PBS.
CCL4/MIP-1 beta
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers