missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CCL4/MIP-1 beta Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 451-MB-010/CF

292.86 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


 View more versions of this product

Product Code. 18328787

This item is not returnable. View return policy

Description

Description

Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CCR5. The ED50 for this effect is 0.75-5 ng/mL.
Specifications

Specifications

Q5QNV9
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
M.W. (theoretical): 7.8 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Ccl4
Recombinant
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Bioactivity
6351
CCL4/MIP-1 beta
E. coli-derived mouse CCL4/MIP-1 beta protein Ala24-Asn92
<0.01 EU / 1 μg of the protein by the LAL method.
Unconjugated
Reconstitute at 100 μg/mL in sterile PBS.
CCL4/MIP-1 beta
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers