Learn More
R&D Systems™ Recombinant Mouse CCL6/C10 (Truncated, aa 42-116) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 3210-C-025
285.09 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18176400
  • 362.00 € / 25µg
Explore more special offers
Description
Description
The Recombinant Mouse CCL6/C10 (Truncated, aa 42-116) Protein is derived from E. coli. The Recombinant Mouse CCL6/C10 (Truncated, aa 42-116) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
8.3 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
20305
CCL6/C10 Truncated, aa 42-116
E. coli-derived mouse CCL6/C10 protein Gly42-Ala116
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.