missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CTSA Protein
A cDNA sequence encoding the CTSA was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Gene ID (Entrez) | 19025 |
---|---|
Name | CTSA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Ctsa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15967638
|
enQuireBio™
QP11558-2ug |
2 μg |
131.00 €
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15947638
|
enQuireBio™
QP11558-10ug |
10 μg |
203.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15957638
|
enQuireBio™
QP11558-1mg |
1 mg |
7540.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
19025 | |
Research Use Only | |
Ctsa | |
Mouse | |
His | |
CTSA protein solution (0.5 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |
CTSA Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
APDQDEIDCL PGLAKQPSFR QYSGYLRASD SKHFHYWFVE SQNDPKNSPV VLWLNGGPGC SSLDGLLTEH GPFLIQPDGV TLEYNPYAWN LIANVLYIES PAGVGFSYSD DKMYVTNDTE VAENNYEALK DFFRLFPEYK DNKLFLTGES YAGIYIPTLA VLVMQDPSMN LQGLAVGNGL ASYEQNDNSL VYFAYYHGLL GNRLWTSLQT HCCAQNKCNF YDNKDPECVN NLLEVSRIVG KSGLNIYNLY APCAGGVPGR HRYEDTLVVQ DFGNIFTRLP LKRRFPEALM RSGDKVRLDP PCTNTTAPSN YLNNPYVRKA LHIPESLPRW DMCNFLVNLQ YRRLYQSMNS QYLKLLSSQK YQILLYNGDV DMACNFMGDE WFVDSLNQKM EVQRRPWLVD YGESGEQVAG FVKECSHITF LTIKGAGHMV PTDKPRAAFT MFSRFLNKEP YVEHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |