missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CXCL13/BLC/BCA-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
476.00 €
Specifications
Gene ID (Entrez) | 55985 |
---|---|
Molecular Weight (g/mol) | 9.8 kDa |
Name | CXCL13/BLC/BCA-1 |
Quantity | 25 μg |
Source | E. coli-derived mouse CXCL13/BLC/BCA-1 protein Ile22-Ala109 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18344817
|
R&D Systems™
470-BC-025 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
476.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18178290
|
R&D Systems™
470-BC-025/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
476.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse CXCL13/BLC/BCA-1 Protein is derived from E. coli. The Recombinant Mouse CXCL13/BLC/BCA-1 Protein has been validated for the following applications: Bioactivity.Specifications
55985 | |
CXCL13/BLC/BCA-1 | |
E. coli-derived mouse CXCL13/BLC/BCA-1 protein Ile22-Ala109 | |
ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1, BCA-1, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLC, BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant) | |
Recombinant |
9.8 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |