Learn More
R&D Systems™ Recombinant Mouse CXCL16 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 503-CX-025
 View more versions of this product
Product Code. 18127140
  • 383.00 € / 25µg
Explore more special offers
Description
Description
The Recombinant Mouse CXCL16 Protein is derived from E. coli. The Recombinant Mouse CXCL16 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
9.9 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
66102
CXCL16 Chemokine Domain
E. coli-derived mouse CXCL16 protein Asn27-Pro114
Unconjugated
>97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.