Learn More
R&D Systems™ Recombinant Mouse Erythropoietin R Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 1390-ER-050
285.09 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18106300
  • 362.00 € / 50µg
Explore more special offers
Description
Description
The Recombinant Mouse Erythropoietin R Fc Chimera Protein is derived from NS0. The Recombinant Mouse Erythropoietin R Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
51.3 kDa (monomer)
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
13857
Erythropoietin R Fc Chimera
Mouse myeloma cell line,NS0-derived mouse Erythropoietin R protein, (N-terminus) Mouse Epo R (Ala25-Pro249) Accession &Num; P14753 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Disulfide-linked homodimer
Recombinant
Product Suggestions