Learn More
R&D Systems™ Recombinant Mouse Fc gamma RIII/CD16 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand:  R&D Systems™ 1960-FC-050
414.68 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18315647
  • 543.00 € / 50µg
Explore more special offers
Description
Description
The Recombinant Mouse Fc gamma RIII/CD16 Protein is derived from NS0. The Recombinant Mouse Fc gamma RIII/CD16 Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
P08508.1
14131
Fc gamma RIII (CD16)
Mouse myeloma cell line,NS0-derived mouse Fc gamma RIII (CD16) protein Ala31-Thr215,with a C-terminal 10-His tag
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Supplied as a 0.2μm filtered solution in PBS.
30-45 kDa, reducing conditions
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, -20 to -70° C under sterile conditions after opening.
Recombinant
Product Suggestions