Learn More
R&D Systems™ Recombinant Mouse FGF-4 (aa 67-202) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 7486-F4-025/CF
285.09 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18107200
  • 373.00 € / 25µg
Explore more special offers
Description
Description
The Recombinant Mouse FGF-4 (aa 67-202) Protein is derived from E. coli. The Recombinant Mouse FGF-4 (aa 67-202) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA.
15.2 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
14175
FGF-4 (aa 67-202)
E. coli-derived mouse FGF-4 protein Ser67-Leu202
Monomer
Recombinant
Product Suggestions