missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse FLRG Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
382.00 €
Specifications
Gene ID (Entrez) | 83554 |
---|---|
Molecular Weight (g/mol) | 26 kDa |
Name | Follistatin-related Gene Protein/FLRG |
Quantity | 25 μg |
Source | Mouse myeloma cell line,NS0-derived mouse Follistatin-related Gene Protein/FLRG protein Val24-Val256,with a C-terminal 6-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18158570
|
R&D Systems™
1255-F3-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
382.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18168500
|
R&D Systems™
1255-F3-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
382.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse FLRG Protein is derived from NS0. The Recombinant Mouse FLRG Protein has been validated for the following applications: Bioactivity.Specifications
83554 | |
Follistatin-related Gene Protein/FLRG | |
Mouse myeloma cell line,NS0-derived mouse Follistatin-related Gene Protein/FLRG protein Val24-Val256,with a C-terminal 6-His tag | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
26 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |