Learn More
R&D Systems™ Recombinant Mouse HGFR/c-MET Fc Chimera (NS0-expressed)
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 7065-ME-100
Product Code. 18186530
  • 394.00 € / 100µg
Explore more special offers
Description
Description
The Recombinant Mouse HGF R/c-MET Fc Chimera (NS0-expressed) is derived from NS0. The Recombinant Mouse HGF R/c-MET Fc Chimera (NS0-expressed) has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
96 & 32 kDa (monomer)
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
17295
HGFR/c-MET
Mouse myeloma cell line,NS0-derived mouse HGF R/c-MET protein, (N-terminus) Mouse HGF R/c-Met (Met1-Asn929) Accession &Num; P16056 DIEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met
Recombinant
Product Suggestions