missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IFN b Protein

A cDNA sequence encoding the IFN b was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12374-1mg

 View more versions of this product

Product Code. 15960209

  • 3915.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

15977
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MINYKQLQLQ ERTNIRKCQE LLEQLNGKIN LTYRADFKIP MEMTEKMQKS YTAFAIQEML QNVFLVFRNN FSSTGWNETI VVRLLDELHQ QTVFLKTVLE EKQEERLTWE MSSTALHLKS YYWRVQRYLK LMKYNSYAWM VVRAEIFRNF LIIRRLTRNF QN
Greater than 95% as determined by Analysis by SDS-PAGE.
IFN b Protein
Research Use Only
Ifnb1
Mouse
Untagged
The IFN-Beta (0.25 mg/ml) contains 20mM HEPES (pH6.0), 0.5M NaCl, 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers