missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-7 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
372.00 € - 1870.00 €
Specifications
Gene ID (Entrez) | 16196 |
---|---|
Molecular Weight (g/mol) | 15 kDa |
Name | IL-7 |
Source | E. coli-derived mouse IL-7 protein Glu26-Ile154,with an N-terminal Met |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
18323178
|
R&D Systems™
407-ML-005/CF |
5 μg | Lyophilized from a 0.2μm filtered solution in PBS. |
372.00 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18148040
|
R&D Systems™
407-ML-005 |
5 μg | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
372.00 €
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18339627
|
R&D Systems™
407-ML-025/CF |
25 μg | Lyophilized from a 0.2μm filtered solution in PBS. |
812.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18336367
|
R&D Systems™
407-ML-025 |
25 μg | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
812.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18348767
|
R&D Systems™
407-ML-200/CF |
200 μg | Lyophilized from a 0.2μm filtered solution in PBS. |
1870.00 €
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18793108
|
R&D Systems™
407-ML-200 |
200 μg | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
1870.00 €
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse IL-7 Protein is derived from E. coli. The Recombinant Mouse IL-7 Protein has been validated for the following applications: Bioactivity.Specifications
16196 | |
IL-7 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
15 kDa | |
E. coli-derived mouse IL-7 protein Glu26-Ile154,with an N-terminal Met | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |