missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Integrin alpha 4 beta 1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 6054-A4-050
346.42 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Mouse Integrin alpha 4 beta 1 Protein is derived from CHO. The Recombinant Mouse Integrin alpha 4 beta 1 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
112.9 kDa (Integrin alpha4) & 86.4 kDa (Integrin beta1) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
16402 | |
Integrin alpha 4 beta 1 | |
Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha 4 beta 1 protein, (N-terminus) Mouse Integrin alpha 4 Tyr34-Gln970 (Arg591Leu) Accession &Num; AAH68313 GGGSGGGSGGGS Acidic Tail 6-His tag Mouse Integrin beta 1 (Gln2-Asp728) Accession &Num; P09055 GGGSGGGSGGGS Basic Tail (C-terminus) | |
CD49d, IA4 | |
Recombinant |