missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Integrin alpha V beta 5 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 7706-AV-050
404.31 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Mouse Integrin alpha V beta 5 Protein is derived from CHO. The Recombinant Mouse Integrin alpha V beta 5 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
Integrin alpha V beta 5 | |
Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha V beta 5 protein, (N-terminus) Mouse Integrin alpha V (Phe31-Val988) Accession &Num; P43406 HP GS Linker Acidic Tail HHHHHH Mouse Integrin beta 5 (Gly24-Asn719) Accession &Num; NP_001139356 HP GS Linker Basic Tail (C-terminus) | |
Noncovalently-linked heterodimer | |
Recombinant |
115 kDa ( alphaV subunit) & 84.8 kDa (beta5 subunit) | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |