missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Lymphotoxin alpha2/beta1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
390.00 €
Specifications
Gene ID (Entrez) | 16992 |
---|---|
Molecular Weight (g/mol) | 18.5 kDa (LT alpha), 18.4 kDa (LTbeta) |
Name | Lymphotoxin α2/β1 |
Quantity | 10 μg |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse Lymphotoxin protein Leu34-Leu202 (LT alpha ) & Gly136-Gly306 (LT beta ) |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18107630
|
R&D Systems™
1008-LY-010/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
390.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse Lymphotoxin alpha2/beta1 Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse Lymphotoxin alpha2/beta1 Protein has been validated for the following applications: Bioactivity.Specifications
16992 | |
Lymphotoxin α2/β1 | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse Lymphotoxin protein Leu34-Leu202 (LT alpha ) & Gly136-Gly306 (LT beta ) | |
Noncovalently-linked heterotrimer | |
Recombinant |
18.5 kDa (LT alpha), 18.4 kDa (LTbeta) | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title