missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse R-Spondin 1 (CHO-expressed) Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

367.00 €

Specifications

Gene ID (Entrez) 192199
Molecular Weight (g/mol) 27 kDa
Name R-Spondin 1
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse R-Spondin 1 protein Met1-Gln265
View More Specs

Products 2
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
18361418
View Documents
R&D Systems™
7150-RS-025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
367.00 €
25µg
Please to purchase this item. Need a web account? Register with us today!
18148400
View Documents
R&D Systems™
7150-RS-025/CF
Lyophilized from a 0.2μm filtered solution in PBS.
367.00 €
25µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Recombinant Mouse R-Spondin 1 (CHO-expressed) Protein is derived from CHO. The Recombinant Mouse R-Spondin 1 (CHO-expressed) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

192199
R-Spondin 1
Chinese Hamster Ovary cell line,CHO-derived mouse R-Spondin 1 protein Met1-Gln265
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
27 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
Videos
SDS
Documents

Documents

Product Certifications