missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Spinesin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 1928-SE-010
315.33 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its ability to cleave the fluorogenic peptide substrate Boc-QAR-AMC (Catalog # ES014 ). The specific activity is >1,000 pmol/min/μg, as measured under the described conditions.Specifications
NP_109634 | |
Supplied as a 0.2 μm filtered solution in MES and NaCl. | |
M.W. (Observed): 55-61 kDa, reducing conditions; M.W. (theoretical): 43 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain |
Enzyme Activity | |
80975 | |
Spinesin | |
Mouse myeloma cell line, NS0-derived mouse Spinesin protein Tyr61-Arg445, with an N-terminal 7-His tag Accession # NP_109634 | |
Pro form | |
Tmprss5 | |
Recombinant | |
Spinesin |