missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse sTNF RI/TNFRSF1A Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

549.00 €

Specifications

Gene ID (Entrez) 21937
Molecular Weight (g/mol) 21 kDa
Name TNF RI/TNFRSF1A
Quantity 50 μg
Source E. coli-derived mouse TNF RI/TNFRSF1A protein Ile22-Ala212,with an N-terminal Met
View More Specs

Products 2
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
18138070
View Documents
R&D Systems™
425-R1-050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
549.00 €
50µg
Please to purchase this item. Need a web account? Register with us today!
18146170
View Documents
R&D Systems™
425-R1-050/CF
Lyophilized from a 0.2μm filtered solution in PBS.
549.00 €
50µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Recombinant Mouse sTNF RI/TNFRSF1A Protein is derived from E. coli. The Recombinant Mouse sTNF RI/TNFRSF1A Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

21937
TNF RI/TNFRSF1A
E. coli-derived mouse TNF RI/TNFRSF1A protein Ile22-Ala212,with an N-terminal Met
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
21 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
Videos
SDS
Documents

Documents

Product Certifications